Skip to Content
Merck
Metalaxyl certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Metalaxyl

Metalaxyl certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-DL-alanine methyl ester
Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade