Mestranol
Synonym(s):
(17α)-3-Methoxy-19-norpregna-1,3,5(10)-trien-20-yn-17-ol, 17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol 3-methyl ether, 17α-Ethynylestradiol 3-methyl ether, 3-Methoxy-17α-ethynylestradiol, NSC 84032
Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing