
Meloxicam

Synonym(s):
Meloxicam, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide
Empirical Formula (Hill Notation):
C14H13N3O4S2
CAS Number:
Molecular Weight:
351.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3