Skip to Content
Merck
Melatonin Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Melatonin Related Compound A

Melatonin Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
5-Methoxytryptamine, 2-(5-Methoxyindol-3-yl)ethylamine, 3-(2-Aminoethyl)-5-methoxyindole, 5-MOT, 5-Methoxyindole-3-ethanamine, Deacetylmelatonin, NSC 56422
Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing