Skip to Content
Merck
Medium 199 With Hanks′ salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Medium 199

Medium 199 With Hanks′ salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Synonym(s):
Medium 199

Products

Product Category
Brand
pH Value
Physical Form
Earles Salts
Hanks Salts
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Sodium Pyruvate (g/L)
Quality Segment
Sterilization Method
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing