Skip to Content
Merck
Mannitol British Pharmacopoeia (BP) Reference Standard

Mannitol

Mannitol British Pharmacopoeia (BP) Reference Standard
Synonym(s):
D-Mannitol, Mannite
Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
EC Number:

Products

Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique