Magenta-Gal
Synonym(s):
Red-Gal, 5-Bromo-6-chloro-3-indolyl β-D-galactopyranoside
Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Products
Brand
Formula Weight
Physical Form
Product Line
Purity
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing