
Luminol sodium salt

Synonym(s):
3-Aminophthalhydrazide, 5-Amino-2,3-dihydro-1,4-phthalazinedione
Linear Formula:
C8H6N3O2Na
CAS Number:
Molecular Weight:
199.14
Products
Product Category
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing