
L-Selectride® solution

Synonym(s):
Lithium tri-sec-butylborohydride solution
Linear Formula:
Li[CH(CH3)CH2CH3]3BH
CAS Number:
Molecular Weight:
190.10
Products
Brand
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing