Lopinavir
Synonym(s):
Lopinavir
Empirical Formula (Hill Notation):
C37H48N4O5
CAS Number:
Molecular Weight:
628.80
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3