Skip to Content
Merck
L-Leucine-1-13C,15N endotoxin tested, 98 atom % 15N, 99 atom % 13C

L-Leucine-1-13C,15N

L-Leucine-1-13C,15N endotoxin tested, 98 atom % 15N, 99 atom % 13C
Linear Formula:
(CH3)2CHCH2CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
133.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2