Skip to Content
Merck
Lithium phosphate monobasic ≥99.99%

Lithium phosphate monobasic

Lithium phosphate monobasic ≥99.99%
Synonym(s):
Lithium dihydrogenphosphate
Linear Formula:
LiH2PO4
CAS Number:
Molecular Weight:
103.93
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing