
Lithium acetate dihydrate

Synonym(s):
Acetic acid lithium salt
Linear Formula:
CH3COOLi · 2H2O
CAS Number:
Molecular Weight:
102.02
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Showing 1-3 of 3