Skip to Content
Merck
Lidocaine Related Compound H Pharmaceutical Secondary Standard; Certified Reference Material

Lidocaine Related Compound H

Lidocaine Related Compound H Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
N-(2,6-Dimethylphenyl)chloroacetamide, 1-Chloroacetylamino-2,6-dimethylbenzene, 2-Chloro-2′,6′-acetoxylidide, 2-Chloro-2′,6′-dimethylacetanilide, 2-Chloro-N-(2,6-dimethylphenyl)acetamide, N-(2,6-Dimethylphenyl)-2-chloroacetamide, N-(2,6-Xylyl)chloroacetamide, N-Chloroacetyl-2,6-dimethylaniline, Chloroacetamido-2,6-xylidine
Linear Formula:
ClCH2CONHC6H3(CH3)2
CAS Number:
Molecular Weight:
197.66
EC Number:

Products

Formula Weight
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1