Levocabastine hydrochloride
Synonym(s):
Levophta, Levostin, R50547, [3S-[1(cis),3α,4β]]-1-[4-cyano-4-(4-fluorophenyl)cyclohexyl]-3-methyl-4-phenyl-4-piperidinecarboxylic acid, Levocabastine Hydrochloride
Empirical Formula (Hill Notation):
C26H30ClFN2O2
CAS Number:
Molecular Weight:
456.98
Products
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2