![Levamisole hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/341/047/536d33e2-b699-4e43-977d-483b74ffb1fc/640/536d33e2-b699-4e43-977d-483b74ffb1fc.png)
Levamisole Hydrochloride
![Levamisole hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/341/047/536d33e2-b699-4e43-977d-483b74ffb1fc/640/536d33e2-b699-4e43-977d-483b74ffb1fc.png)
Synonym(s):
(−)-Tetramisole hydrochloride, (S)-(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Levamisole hydrochloride, (−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo(2,1-b)thiazole, S(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride
Empirical Formula (Hill Notation):
C11H12N2S · HCl
CAS Number:
Molecular Weight:
240.75
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3