Skip to Content
Merck
L-Cystine dihydrochloride, non-animal derived

L-Cystine dihydrochloride, non-animal derived

L-Cystine dihydrochloride, non-animal derived
Synonym(s):
L-Cystine dihydrochloride
Empirical Formula (Hill Notation):
C6H12N2O4S2 · 2HCl
CAS Number:
Molecular Weight:
313.22
EC Number:

Products

Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing