Skip to Content
Merck
Lanthanum(III) chloride hydrate 99.9% trace metals basis

Lanthanum(III) chloride hydrate

Lanthanum(III) chloride hydrate 99.9% trace metals basis
Linear Formula:
LaCl3 · xH2O
CAS Number:
Molecular Weight:
245.26 (anhydrous basis)

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing