Isomalt
Synonym(s):
Isomalt, 6-O-α-D-Glucopyranosyl-D-glucitol mixed with 1-O-α-D-glucopyranosyl-D-mannitol
Empirical Formula (Hill Notation):
C12H24O11
CAS Number:
Molecular Weight:
344.31
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing