![Iriflophenone 3-C-glucoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/329/093/53826f71-d051-49d2-b0d4-7ffa601227ed/640/53826f71-d051-49d2-b0d4-7ffa601227ed.png)
Iriflophenone 3-C-glucoside
![Iriflophenone 3-C-glucoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/329/093/53826f71-d051-49d2-b0d4-7ffa601227ed/640/53826f71-d051-49d2-b0d4-7ffa601227ed.png)
Synonym(s):
Iriflophenone 3-C-β-D-glucopyranoside, (3-β-D-Glucopyranosyl-2,4,6-trihydroxyphenyl)(4-hydroxyphenyl)methanone, Iriflophenone 3-C-β-D-glucoside
Empirical Formula (Hill Notation):
C19H20O10
CAS Number:
Molecular Weight:
408.36
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing