Iotrolan for system suitability
Synonym(s):
Iotrolan
Empirical Formula (Hill Notation):
C37H48I6N6O18
CAS Number:
Molecular Weight:
1626.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing