Skip to Content
Merck
Imipenem monohydrate ≥98% (HPLC)

Imipenem monohydrate

Imipenem monohydrate ≥98% (HPLC)
Synonym(s):
(5R,6S)-6-[(1R)-1-Hydroxyethyl]-3-[[2-[(iminomethyl)amino]ethyl]thio]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid monohydrate, Primaxin monohydrate, Tienam monohydrate
Empirical Formula (Hill Notation):
C12H17N3O4S · H2O
CAS Number:
Molecular Weight:
317.36
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing