![Imidazole hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/321/525/04283f83-5c42-4256-b351-9b0106e0105c/640/04283f83-5c42-4256-b351-9b0106e0105c.png)
Imidazole hydrochloride
![Imidazole hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/321/525/04283f83-5c42-4256-b351-9b0106e0105c/640/04283f83-5c42-4256-b351-9b0106e0105c.png)
Synonym(s):
Imidazole hydrochloride
Empirical Formula (Hill Notation):
C3H4N2 · HCl
CAS Number:
Molecular Weight:
104.54
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing