![Imazamethabenz-methyl certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/309/081/708dceea-8fb1-4408-9ebf-8c75420c1b70/640/708dceea-8fb1-4408-9ebf-8c75420c1b70.png)
Imazamethabenz-methyl
![Imazamethabenz-methyl certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/309/081/708dceea-8fb1-4408-9ebf-8c75420c1b70/640/708dceea-8fb1-4408-9ebf-8c75420c1b70.png)
Empirical Formula (Hill Notation):
C16H20N2O3
CAS Number:
Molecular Weight:
288.34
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing