Idoxuridine
Synonym(s):
5-Iodo-2′-deoxyuridine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-iodouracil, 2′-Deoxy-5-iodouridine, 5-IUdR, IDU, IdUrd, Idoxuridine
Empirical Formula (Hill Notation):
C9H11IN2O5
CAS Number:
Molecular Weight:
354.10
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3