IBUPROFEN IMPURITY L
Synonym(s):
1-Hydroxyibuprofen, 2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propionic acid
Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28
Products
Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing