
Hydroxyzine Related Compound A

Synonym(s):
1-(4-Chlorobenzhydryl)piperazine, (RS)-1-[(4-Chlorophenyl)phenylmethyl]piperazine, NSC 86164, Norchlorcyclizine
Empirical Formula (Hill Notation):
C17H19ClN2
CAS Number:
Molecular Weight:
286.80
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing