Skip to Content
Merck
Hydroxychloroquine sulfate European Pharmacopoeia (EP) Reference Standard

Hydroxychloroquine sulfate

Hydroxychloroquine sulfate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Hydroxychloroquine sulfate, 7-Chloro-4-[4-(N-ethyl-N-b-hydroxyethylamino)-1-methylbutylamino]quinoline sulfate, Hydroxychloroquine Sulfate
Empirical Formula (Hill Notation):
C18H26ClN3O · H2SO4
CAS Number:
Molecular Weight:
433.95
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique