
Hydroxycarbamide

Synonym(s):
Hydroxyurea, Hydroxycarbamide
Linear Formula:
NH2CONHOH
CAS Number:
Molecular Weight:
76.05
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing