![Hydrazinium dichloride for synthesis](/deepweb/assets/sigmaaldrich/product/images/299/384/81fbef1e-554f-4957-886b-81820118ef0b/640/81fbef1e-554f-4957-886b-81820118ef0b.jpg)
Hydrazinium dichloride
![Hydrazinium dichloride for synthesis](/deepweb/assets/sigmaaldrich/product/images/299/384/81fbef1e-554f-4957-886b-81820118ef0b/640/81fbef1e-554f-4957-886b-81820118ef0b.jpg)
Synonym(s):
Hydrazinium dichloride
Empirical Formula (Hill Notation):
H4N2 · 2HCl
CAS Number:
Molecular Weight:
104.97
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing