
Hydralazine hydrochloride

Synonym(s):
1-Hydrazinophthalazine hydrochloride, Hydralazine hydrochloride
Empirical Formula (Hill Notation):
C8H8N4 · HCl
CAS Number:
Molecular Weight:
196.64
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3