![Hordenine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/292/139/474a95e7-f08d-4938-a7fe-d8d34f1a0c75/640/474a95e7-f08d-4938-a7fe-d8d34f1a0c75.png)
Hordenine
![Hordenine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/292/139/474a95e7-f08d-4938-a7fe-d8d34f1a0c75/640/474a95e7-f08d-4938-a7fe-d8d34f1a0c75.png)
Synonym(s):
2-(4-Hydroxyphenyl)-N,N-dimethylethylamine, 4-(2-Dimethylaminoethyl)phenol, N,N-Dimethyltyramine, p-Hydroxy-N,N-dimethylphenethylamine, Anhaline, Cactine, Eremursin, Peyocactine
Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing