Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
High permeability magnetic alloy, Ni80Fe15Mo5
Products
Manufacturer
Physical Form
Diameter (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF51900027
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 6mm disks, 0.012mm thickness
Expand
View Pricing
GF87502877
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 8mm disks, 0.012mm thickness
Expand
View Pricing
GF98455832
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 50 x 50mm, 0.012mm thickness, light tight (lt)
Expand
View Pricing
GF37859305
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 10mm disks, 0.012mm thickness
Expand
View Pricing
GF06979326
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 100 x 100mm, 0.012mm thickness, light tight (lt)
Expand
View Pricing
GF72415201
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 4mm disks, 0.012mm thickness
Expand
View Pricing
GF60787648
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 25 x 25mm, 0.012mm thickness, light tight (lt)
Expand
View Pricing
GF32413025
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 15mm disks, 0.012mm thickness
Expand
View Pricing
GF51628226
High permeability magnetic alloy, Ni80Fe15Mo5,
foil, 25mm disks, 0.012mm thickness
Expand
View Pricing