![Hexetidine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/183/761/62ac2398-c71c-49cd-b7f3-597022373396/640/62ac2398-c71c-49cd-b7f3-597022373396.png)
Hexetidine
![Hexetidine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/183/761/62ac2398-c71c-49cd-b7f3-597022373396/640/62ac2398-c71c-49cd-b7f3-597022373396.png)
Synonym(s):
Hexetidine, mixture of stereoisomers, 5-Amino-1,3-bis(2-ethylhexyl)hexahydro-5-methylpyrimidine, NSC 17764
Empirical Formula (Hill Notation):
C21H45N3
CAS Number:
Molecular Weight:
339.60
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing