Skip to Content
Merck
Hexadecyltrimethylammonium hydroxide solution ~25% in methanol (T)

Hexadecyltrimethylammonium hydroxide solution

Hexadecyltrimethylammonium hydroxide solution ~25% in methanol (T)
Linear Formula:
CH3(CH2)15N(OH)(CH3)3
CAS Number:
Molecular Weight:
301.55

Products

Product Category
Brand
Formula Weight
Physical Form
Special Grade