![Hexadecanedioic acid 96%](/deepweb/assets/sigmaaldrich/product/structures/924/074/79be8b12-288d-4aa2-81f3-2a5aba819e0d/640/79be8b12-288d-4aa2-81f3-2a5aba819e0d.png)
Hexadecanedioic acid
![Hexadecanedioic acid 96%](/deepweb/assets/sigmaaldrich/product/structures/924/074/79be8b12-288d-4aa2-81f3-2a5aba819e0d/640/79be8b12-288d-4aa2-81f3-2a5aba819e0d.png)
Synonym(s):
Thapsic acid, Hexadecanedioic acid
Linear Formula:
HOOC(CH2)14COOH
CAS Number:
Molecular Weight:
286.41
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing