![Hexaconazol PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/265/605/45a4cf1b-9bf7-4dd8-a49e-33a6c91c7314/640/45a4cf1b-9bf7-4dd8-a49e-33a6c91c7314.png)
Hexaconazol
![Hexaconazol PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/265/605/45a4cf1b-9bf7-4dd8-a49e-33a6c91c7314/640/45a4cf1b-9bf7-4dd8-a49e-33a6c91c7314.png)
Synonym(s):
(±)-2-(2,4-Dichlorophenyl)-1-(1H-1,2,4-triazol-1-yl)-2-hexanol
Empirical Formula (Hill Notation):
C14H17Cl2N3O
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2