Skip to Content
Merck
Hexachlorophene ≥98% (HPLC)

Hexachlorophene

Hexachlorophene ≥98% (HPLC)
Synonym(s):
2,2′-Methylenebis(3,4,6-trichlorophenol), 2,2′,3,3′,5,5′-Hexachloro-6,6′-dihydroxydiphenylmethane, 2,2′-Dihydroxy-3,3′,5,5′,6,6′-hexachlorodiphenylmethane, 2,2′-Methylenebis[3,4,6-trichlorophenol], Nabac
Linear Formula:
CH2[C6H(Cl)3OH]2
CAS Number:
Molecular Weight:
406.90

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing