Skip to Content
Merck
Haloxyfop-2-ethoxyethyl PESTANAL®, analytical standard

Haloxyfop-2-ethoxyethyl

Haloxyfop-2-ethoxyethyl PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C19H19ClF3NO5
CAS Number:
Molecular Weight:
433.81
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing