Skip to Content
Merck
H-7 dihydrochloride ≥98% (HPLC)

H-7 dihydrochloride

H-7 dihydrochloride ≥98% (HPLC)
Synonym(s):
1-(5-Isoquinolinylsulfonyl)-2-methylpiperazine dihydrochloride, Isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride
Empirical Formula (Hill Notation):
C14H17N3O2S · 2HCl
CAS Number:
Molecular Weight:
364.29

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing