
Guanethidine Monosulfate

Synonym(s):
Guanethidine monosulfate (1:1), 1-(2-Guanidinoethyl)octahydroazocine
Linear Formula:
C10H22N4 · H2SO4
CAS Number:
Molecular Weight:
296.39
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2