Guaiacol
Synonym(s):
2-Methoxyphenol, Catechol monomethyl ether, Pyrocatechol monomethyl ether
Linear Formula:
(CH3O)C6H4OH
CAS Number:
Molecular Weight:
124.14
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4