Skip to Content
Merck
Glutaric anhydride 95%

Glutaric anhydride

Glutaric anhydride 95%
Synonym(s):
Glutaric anhydride
Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing