
Glimepiride Related Compound D

Synonym(s):
3-Ethyl-2,5-dihydro-4-methyl-N-[2-[3-[[[[(trans-4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-oxo-1H-pyrrole-1-carboxamide, 1-[[m-[2-(3-Ethyl-4-methyl-2-oxo-3-pyrroline-1-carboxamido)ethyl]phenyl]sulfonyl]-3-(trans-4-methylcyclohexyl)urea
Empirical Formula (Hill Notation):
C24H34N4O5S
CAS Number:
Molecular Weight:
490.62
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing