
Ginkgolide B

Empirical Formula (Hill Notation):
C20H24O10
CAS Number:
Molecular Weight:
424.40
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing