Skip to Content
Merck
Geranyl tiglate ≥95%, FG

Geranyl tiglate

Geranyl tiglate ≥95%, FG
Empirical Formula (Hill Notation):
C15H24O2
CAS Number:
Molecular Weight:
236.35

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing