Skip to Content
Merck
Frovatriptan succinate monohydrate ≥97% (HPLC)

Frovatriptan succinate monohydrate

Frovatriptan succinate monohydrate ≥97% (HPLC)
Synonym(s):
Frovatriptan succinate, R-(+) 3-Methylamino-6-carboxamido-1,2,3,4-tetrahydrocarbazole monosuccinate monohydrate
Empirical Formula (Hill Notation):
C14H17N3O · C4H6O4 · H2O
CAS Number:
Molecular Weight:
379.41

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing