Skip to Content
Merck
Fluorouracil Related Compound E certified reference material, pharmaceutical secondary standard

Fluorouracil Related Compound E

Fluorouracil Related Compound E certified reference material, pharmaceutical secondary standard
Synonym(s):
5-Chlorouracil, 5-Chloro-2,4-dihydroxypyrimidine, 5-Chloropyrimidine-2,4(1H,3H)-dione
Empirical Formula (Hill Notation):
C4H3ClN2O2
CAS Number:
Molecular Weight:
146.53

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1