Skip to Content
Merck
Fluoroquinolonic acid Pharmaceutical Secondary Standard; Certified Reference Material

Fluoroquinolonic acid

Fluoroquinolonic acid Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid, Fluoroquinolonic acid, Q-Acid
Empirical Formula (Hill Notation):
C13H9ClFNO3
CAS Number:
Molecular Weight:
281.67
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing