
Flunixin Related Compound C

Synonym(s):
Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-chloronicotinate
Empirical Formula (Hill Notation):
C8H8ClNO2
CAS Number:
Molecular Weight:
185.61
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing